Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC22A14 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569268
Description
This target displays homology in the following species: Cow: 79%; Dog: 79%; Guinea Pig: 79%; Horse: 86%; Human: 100%; Mouse: 86%; Pig: 86%; Rat: 86%.
SLC22A14 is a protein coding gene.
Specifications
SLC22A14 | |
Polyclonal | |
Unconjugated | |
SLC22A14 | |
Gm1128; LOW QUALITY PROTEIN: solute carrier family 22 member 14; OCTL2; OCTL4; ORCTL4; ORCTL-4; organic cation transporter like 4; organic cation transporter-like 4; organic cationic transporter-like 4; SLC22A14; solute carrier family 22 (organic cation transporter), member 14; solute carrier family 22 member 14; solute carrier family 22, member 14 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
9389 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9Y267 | |
SLC22A14 | |
synthetic peptide directed towards the N-terminal region of human SLC22A14. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction