Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC22A16 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP185410
Description
SLC22A16 Polyclonal specifically detects SLC22A16 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SLC22A16 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
SLC22A16 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:KLPETLGKRLATTWEEAAKLESENESKSSKLLLTTNNSGLEKTEAITPRDSGLGE | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
0.05mg/mL | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Carnitine transporter 2, CT2Organic cation/carnitine transporter 6, Flipt 2, FLIPT2member 16, fly-like putative organic ion transporter 2, Fly-like putative transporter 2, organic cation transporter 6, Organic cation transporter OKB1, solute carrier family 22 (organic cation/carnitine transporter), member 16, solute carrier family 22 member 16 | |
Rabbit | |
Affinity Purified | |
RUO | |
85413 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction