Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC22A16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142244
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142244 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142244 Supplier Invitrogen™ Supplier No. PA5142244
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.

This gene encodes a member of the organic zwitterion transporter protein family which transports carnitine. The encoded protein has also been shown to transport anticancer drugs like bleomycin successful treatment has been correlated with the level of activity of this transporter in tumor cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC22A16
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene SLC22A16
Gene Accession No. Q86VW1
Gene Alias Carnitine transporter 2; CT2; dJ261K5.1; epididymis secretory protein Li 18; flipt 2; FLIPT2; fly-like putative organic ion transporter 2; fly-like putative transporter 2; HEL-S-18; OAT6; OCT6; OKB1; organic cation transporter 6; organic cation transporter OKB1; Organic cation/carnitine transporter 6; SLC22A16; solute carrier family 22 (organic cation transporter), member 16; solute carrier family 22 (organic cation/carnitine transporter), member 16; solute carrier family 22 member 16; WUGSC:RG331P03.1
Gene Symbols SLC22A16
Host Species Goat
Immunogen Peptide with sequence CSRNKRENTSSL.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85413
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.