Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC22A17 Monoclonal Antibody (OTI2D6)
GREENER_CHOICE

Catalog No. PIMA527266
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527266 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527266 Supplier Invitrogen™ Supplier No. MA527266
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SLC22A17 Monoclonal Antibody for Western Blot, IHC (P)

The Slc22 family of organic anion and cation transporters (OATs, OCTs, OCTNs) are transmembrane proteins expressed predominantly in kidney and liver. Each contain 12 predicted alpha-helical transmembrane domains (TMDs) and one large extracellular loop between TMDs 1 and 2. Transporters of the SLC22 family function in different ways such as uniporters that mediate facilitated diffusion in either direction (OCTs), as anion exchangers (OAT1, OAT3 and URAT1), and as Na(+)/l-carnitine cotransporter (OCTN2). Slc22 family members participate in the absorption and/or excretion of drugs, xenobiotics, and endogenous compounds in intestine, liver, and kidney, and perform homeostatic functions in brain and heart. Mutations in the Slc22 family may cause specific diseases such as primary systemic carnitine deficiency or idiopathic renal hypouricemia and may change drug absorption or excretion. Recent studies show the expression of Slc22A17 as receptor for Lipocalin 2 is relatively high in hematopoietic stem cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC22A17
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2D6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SLC22A17
Gene Accession No. Q8WUG5
Gene Alias 1700094C23Rik; 24p3 receptor; 24p3R; AU041908; AW555662; BOCT; Boit; brain-specific organic ion transporter; brain-type organic cation transporter; hBOIT; Lipocalin-2 receptor; mBOCT; Neutrophil gelatinase-associated lipocalin receptor; NGAL receptor; NgalR; NGALR2; NGALR3; organic cation transporter BOCT; potent brain type organic ion transporter; rBOCT; SLC22A17; solute carrier family 22 (organic cation transporter), member 17; solute carrier family 22 member 17; solute carrier family 22, member 17
Gene Symbols SLC22A17
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 241-300 of SLC22A17 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51310
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.