Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC22A2 Monoclonal Antibody (640438)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA524289

Catalog No. PIMA524289


Only null left
Add to Cart

Description

Description

Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.

Organic cation transporters (OCT) are expressed in the plasma membrane of epithelial cells from a wide range of tissues, where they function in the elimination of endogenous amines, cationic drugs and other xenobiotics. The structure of OCTs consists of a 12-transmembrane-domain structure and a large extracellular hydrophilic loop. In humans, OCT1 is primarily expressed in the liver, while OCT2 is expressed in the kidney. OCT3 is expressed in the placenta, skeletal muscle, prostate, aorta and liver. OCT2, also known as SLC22A2, is a multi-specific transporter protein localizing to the basolateral and luminal membranes of the kidney distal tubule and proximal tubules. OCT2 is responsible for mediating the pH-sensitive tubular uptake of organic compounds from circulation. An additional splice variant exists for OCT2, namely OCT2-A.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SLC22A2
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
O15244
Slc22a2
NS0 mouse myeloma cell line transfected with human SLC-22A2.
100 μg
Primary
Human
Antibody
IgG2a
Flow Cytometry, Immunohistochemistry (Paraffin)
640438
Unconjugated
Slc22a2
2-Oct; hOCT2; OCT2; OCT2r; Orct2; Organic cation transporter 2; rOCT2; Slc22a2; solute carrier family 22 (organic cation transporter), member 2; solute carrier family 22 member 2; solute carrier family 22, member 2
Mouse
Protein A/G
RUO
6582
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.