Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC22A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552579
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552579 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552579 Supplier Invitrogen™ Supplier No. PA552579
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ESPRWLISQN KNAEAMRIIK HIAKKNGKSL PASLQRLRLE EETGKKLNPS FLDLVRTPQI R Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 77%.

Organic cation transporters (OCT) are expressed in the plasma membrane of epithelial cells from a wide range of tissues, where they function in the elimination of endogenous amines, cationic drugs and other xenobiotics. The structure of OCTs consists of a 12-transmembrane-domain structure and a large extracellular hydrophilic loop. In humans, OCT1 is primarily expressed in the liver, while OCT2 is expressed in the kidney. OCT3 is expressed in the placenta, skeletal muscle, prostate, aorta and liver. OCT2, also known as SLC22A2, is a multi-specific transporter protein localizing to the basolateral and luminal membranes of the kidney distal tubule and proximal tubules. OCT2 is responsible for mediating the pH-sensitive tubular uptake of organic compounds from circulation. An additional splice variant exists for OCT2, namely OCT2-A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC22A2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Slc22a2
Gene Accession No. O15244
Gene Alias 2-Oct; hOCT2; OCT2; OCT2r; Orct2; Organic cation transporter 2; rOCT2; Slc22a2; solute carrier family 22 (organic cation transporter), member 2; solute carrier family 22 member 2; solute carrier family 22, member 2
Gene Symbols Slc22a2
Host Species Rabbit
Immunogen Recombinant protein fragment of Human SLC22A2 (Product #RP-101762)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6582
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.