Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC22A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112715
Description
SLC22A2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Organic cation transporters (OCT) are expressed in the plasma membrane of epithelial cells from a wide range of tissues, where they function in the elimination of endogenous amines, cationic drugs and other xenobiotics. The structure of OCTs consists of a 12-transmembrane-domain structure and a large extracellular hydrophilic loop. In humans, OCT1 is primarily expressed in the liver, while OCT2 is expressed in the kidney. OCT3 is expressed in the placenta, skeletal muscle, prostate, aorta and liver. OCT2, also known as SLC22A2, is a multi-specific transporter protein localizing to the basolateral and luminal membranes of the kidney distal tubule and proximal tubules. OCT2 is responsible for mediating the pH-sensitive tubular uptake of organic compounds from circulation. An additional splice variant exists for OCT2, namely OCT2-A.
Specifications
| SLC22A2 | |
| Polyclonal | |
| Unconjugated | |
| Slc22a2 | |
| 2-Oct; hOCT2; OCT2; OCT2r; Orct2; Organic cation transporter 2; rOCT2; Slc22a2; solute carrier family 22 (organic cation transporter), member 2; solute carrier family 22 member 2; solute carrier family 22, member 2 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 20518, 29503, 6582 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| O15244, O70577, Q9R0W2 | |
| Slc22a2 | |
| Recombinant Human Solute carrier family 22 member 2 protein (48-135AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction