Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC22A24 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560895
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560895 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560895 Supplier Invitrogen™ Supplier No. PA560895
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PVSEAMVDIE RKIVTPGICS VSGLVLSHDV H Highest antigen sequence identity to the following orthologs: Mouse - 39%, Rat - 39%.

SLC22A24 belongs to a large family of transmembrane proteins that function as uniporters, symporters, and antiporters to transport organic ions across cell membranes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC22A24
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC22A24
Gene Accession No. Q8N4F4
Gene Alias NET46; SLC22A24; Solute carrier family 22 member 24; solute carrier family 22, member 24
Gene Symbols SLC22A24
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC22A24. Recombinant protein control fragment (Product #RP-100478).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283238
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.