Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC22A25 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113010
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113010 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113010 Supplier Invitrogen™ Supplier No. PA5113010
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC22A25 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

SLC22A25 gene ontology annotations related to this gene include transmembrane transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC22A25
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.44 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SLC22A25
Gene Accession No. Q6T423
Gene Alias HIMTP; integral membrane transport UST1r; MGI:2442751, MGI:2385316, MGI:3042283, MGI:3645714, MGI:3605624, MGI:2442750; organic anion transporter UST6; putative integral membrane transport UST1r; putative UST1-like organic anion transporter; SLC22A25; Slc22a9; solute carrier family 22 (organic anion/cation transporter), member 9; solute carrier family 22 member 24; solute carrier family 22 member 25; solute carrier family 22, member 25; Ust1; Ust1r; UST6
Gene Symbols SLC22A25
Host Species Rabbit
Immunogen Recombinant Human Solute carrier family 22 member 25 protein (31-145AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387601
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.