Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC22A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542486
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542486 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542486 Supplier Invitrogen™ Supplier No. PA542486
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PDHYWCRGPS AAALAERCGW SPEEEWNRTA PASRGPEPPE RRGRCQRYLL Sequence homology: Cow: 79%; Human: 100%; Pig: 86%.

Polyspecific organic cation transporters in the liver, kidney, intestine, and other organs are critical for elimination of many endogenous small organic cations as well as a wide array of drugs and environmental toxins. This gene is one of three similar cation transporter genes located in a cluster on chromosome 6. The encoded protein contains twelve putative transmembrane domains and is a plasma integral membrane protein. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC22A3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC22A3
Gene Accession No. O75751
Gene Alias 3-Oct; EMT; EMT organic cation transporter 3; EMTH; extraneuronal monoamine extraneuronal monoamine; extraneuronal monoamine transporter; Oct3; Orct3; Organic cation transporter 3; Slc22a3; Slca22a3; solute carrier family 22 (extraneuronal monoamine transporter), member 3; solute carrier family 22 (organic cation transporter), member 3; solute carrier family 22 member 3; solute carrier family 22, member 3
Gene Symbols SLC22A3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SLC22A3 (aa 46-95).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6581
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.