Learn More
Invitrogen™ SLC22A4 Polyclonal Antibody

Description
Positive Samples: SW480, HL-60, NCI-H460, LO2, HeLa, Mouse kidney, Mouse liver, Rat liver; Cellular Location: Membrane, Multi-pass membrane protein.
Specifications
Specifications
| Antigen | SLC22A4 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.74 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 |
| Gene | SLC22A4 |
| Gene Accession No. | Q9H015, Q9R141, Q9Z306 |
| Gene Alias | ergothioneine transporter; ET transporter; ETT; integral membrane transport protein; LSTP-Like 2; Octn1; organic cation transporter 1; organic cation transporter OCTN1; organic cation/carnitine transporter 1; SLC22A4; solute carrier family (organic cation transporter), member 4; solute carrier family 22 (organic cation transporter), member 4; solute carrier family 22 (organic cation/ergothioneine transporter), member 4; solute carrier family 22 (organic cation/zwitterion transporter), member 4; solute carrier family 22 member 4; UT2H |
| Gene Symbols | SLC22A4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.