Learn More
Invitrogen™ SLC22A5 Polyclonal Antibody

Description
Antibody detects endogenous levels of total SLC22A5.
Specifications
Specifications
| Antigen | SLC22A5 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | SLC22A5 |
| Gene Accession No. | O70594, O76082, Q9Z0E8 |
| Gene Alias | CDSP; CT1; high-affinity carnitine transporter; high-affinity sodium dependent carnitine cotransporter; high-affinity sodium-dependent carnitine cotransporter; integral membrane transport protein; juvenile visceral steatosis; jvs; Lstpl; Octn2; OCTN2VT; Organic cation/carnitine transporter 2; S22A5; SLC22A5; solute carrier family 22 (organic cation transporter), member 5; solute carrier family 22 (organic cation/carnitine transporter), member 5; solute carrier family 22 member 5; solute carrier family 22, member 5; UST2r |
| Gene Symbols | SLC22A5 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.