Learn More
SLC22A6 Polyclonal Antibody

Description
SLC22A6 Polyclonal Antibody for Western Blot, IHC (PFA)
Specifications
Specifications
| Antigen | SLC22A6 |
| Applications | Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide |
| Gene | SLC22A6 |
| Gene Accession No. | O35956, Q4U2R8, Q8VC69 |
| Gene Alias | hOAT1; hPAHT; hROAT1; hypothetical protein zgc:77073; kidney-specific transport protein; mNKT; mOat1; mROAT1; NKT; Novel kidney transcript; oat; OAT1; oat3; Orctl1; Organic anion transporter 1; organic cationic transporter-like 1; PAH transporter; PAHT; para-aminohippurate transporter; rbROAT1; Renal organic anion transporter 1; renal organic anion transporter 1 (rbOAT1); ROAT1; rROAT1; slc22a6; slc22a6l; solute carrier family 22 (organic anion transporter), member 6; solute carrier family 22 (organic anion transporter), member 6, like; solute carrier family 22 member 6; solute carrier family 22 member 6 isoform b; solute carrier family 22 member 6, like; solute carrier family 22, member 6; zgc:77073 |
| Gene Symbols | SLC22A6 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.