Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC22A6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102488
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102488 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102488 Supplier Invitrogen™ Supplier No. PA5102488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SLC22A6.

The protein is involved in the sodium-dependent transport and excretion of organic anions, some of which are potentially toxic. The encoded protein is an integral membrane protein and may be localized to the basolateral membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC22A6
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SLC22A6
Gene Accession No. O35956, Q4U2R8, Q8VC69
Gene Alias hOAT1; hPAHT; hROAT1; hypothetical protein zgc:77073; kidney-specific transport protein; mNKT; mOat1; mROAT1; NKT; Novel kidney transcript; oat; OAT1; oat3; Orctl1; Organic anion transporter 1; organic cationic transporter-like 1; PAH transporter; PAHT; para-aminohippurate transporter; rbROAT1; Renal organic anion transporter 1; renal organic anion transporter 1 (rbOAT1); ROAT1; rROAT1; slc22a6; slc22a6l; solute carrier family 22 (organic anion transporter), member 6; solute carrier family 22 (organic anion transporter), member 6, like; solute carrier family 22 member 6; solute carrier family 22 member 6 isoform b; solute carrier family 22 member 6, like; solute carrier family 22, member 6; zgc:77073
Gene Symbols SLC22A6
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLC22A6(Accession Q4U2R8), corresponding to amino acid residues I532-L563.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18399, 29509, 9356
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.