Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC22A7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | SLC22A7 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
SLC22A7 Polyclonal specifically detects SLC22A7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SLC22A7 | |
Polyclonal | |
Purified | |
RUO | |
hOAT2, liver-specific transporter, NLTMGC45202, Novel liver transporter, OAT2MGC24091, Organic anion transporter 2, solute carrier family 22 (organic anion transporter), member 7, solute carrier family 22 member 7 | |
SLC22A7 | |
Primary | |
60 kDa |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Q5T048 | |
10864 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title