Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC22A7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590248
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590248 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590248 Supplier Invitrogen™ Supplier No. PA590248
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ALPGAPANFS HQDVWLEAHL PREPDGTLSS CLRFAYPQAL PNTTLGEERQ SRGELEDEPA TVPCSQGWEY DHSEFSSTIA TESQWDLVCE QKG; Positive Samples: U-251MG, HT-29, mouse liver, mouse kidney, rat kidney; Cellular Location: Basolateral cell membrane, Multi-pass membrane protein.

SLC22A7 is involved in the sodium-independent transport and excretion of organic anions, some of which are potentially toxic. The protein is an integral membrane protein and appears to be localized to the basolateral membrane of the kidney. The protein encoded by this gene is involved in the sodium-independent transport and excretion of organic anions, some of which are potentially toxic. The encoded protein is an integral membrane protein and appears to be localized to the basolateral membrane of the kidney. Alternatively spliced transcript variants encoding different isoforms have been described. The protein encoded by this gene is involved in the sodium-independent transport and excretion of organic anions, some of which are potentially toxic. The encoded protein is an integral membrane protein and appears to be localized to the basolateral membrane of the kidney. Alternatively spliced transcript variants encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC22A7
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SLC22A7
Gene Accession No. Q5RLM2, Q91WU2, Q9Y694
Gene Alias hOAT2; liver-specific transporter; MGC24091; MGC45202; mOAT2; NLT; novel liver transporter; oat2; organic anion transporter 2; rbOAT2; renal organic anion transporter 2; Slc22a7; solute carrier family 22 (organic anion transporter), member 7; solute carrier family 22 member 7; solute carrier family 22 member 7; LOW QUALITY PROTEIN: solute carrier family 22 member 7
Gene Symbols SLC22A7
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108114, 10864, 89776
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.