Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC22A8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120857
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120857 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120857 Supplier Invitrogen™ Supplier No. PA5120857
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: 293T, U-251MG. The target is usually found in the following locations: Basolateral cell membrane, Multi-pass membrane protein. Immunogen sequence: QTGMGVSNLW TRVGSMVSPL VKITGEVQPF IPNIIYGITA LLGGSAALFL PETLNQPLPE TIEDLENWSL RAKKPKQEPE VEKASQRIPL QPHGPGLGSS.

SLC22A8 is involved in the sodium-independent transport and excretion of organic anions, some of which are potentially toxic. SLC22A8 is an integral membrane protein and appears to be localized to the basolateral membrane of the kidney.The protein encoded by this gene is involved in the sodium-independent transport and excretion of organic anions, some of which are potentially toxic. The encoded protein is an integral membrane protein and appears to be localized to the basolateral membrane of the kidney. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC22A8
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.04 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SLC22A8
Gene Accession No. Q8TCC7
Gene Alias hOAT3; MGC24086; mOat3; OAT3; OCT3; organic anion transporter 3; rbOAT3; reduced in osteosclerosis transporter; renal organic anion transporter 3; rOat3; Roct; Slc22a8; solute carrier family 22 (organic anion transporter), member 8; solute carrier family 22 ,member 8; solute carrier family 22 member 8; solute carrier family 22, member 8
Gene Symbols SLC22A8
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 443-542 of human OAT3/SLC22A8 (NP_0042452).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9376
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.