Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC22A9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570675
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570675 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570675 Supplier Invitrogen™ Supplier No. PA570675
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Human: 100%.

Sodium-independent organic anion transporter which exhibits high specificity for sulfated conjugates of xenobiotics and steroid hormones. It is also specifically activated by 3 to 5 carbons-containing short-chain fatty acids/SCFAs, including propionate, butyrate and valerate. May operate the exchange of sulfated organic components against short-chain fatty acids/SCFAs at the sinusoidal membrane of hepatocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC22A9
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC22A9
Gene Accession No. Q8IVM8
Gene Alias hOAT4; OAT4; OAT7; organic anion transporter 4; organic anion transporter 7; SLC22A9; solute carrier family 22 (organic anion transporter), member 9; solute carrier family 22 (organic anion/cation transporter), member 9; solute carrier family 22 member 9; UST3; UST3H
Gene Symbols SLC22A9
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human SLC22A9 (aa 501-550).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114571
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.