Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC23A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561555
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561555 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561555 Supplier Invitrogen™ Supplier No. PA561555
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NTVPGSPEER GLIQWKAGAH ANSDMSSSLK SYDFPIGMGI VKRITFLKYI PICPVFKGFS SSSKDQIAIP EDTPENTETA S Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 74%.

Sodium/ascorbate cotransporter. Mediates electrogenic uptake of vitamin C, with a stoichiometry of 2 Na(+) for each ascorbate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC23A1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC23A1
Gene Accession No. Q9UHI7
Gene Alias D18Ucla2; hSVCT1; Na(+)/L-ascorbic acid transporter 1; SLC23A1; Slc23a2; Sodium-dependent vitamin C transporter 1; sodium-dependent vitamin C transporter-1; solute carrier family 23 (ascorbic acid transporter), member 1; solute carrier family 23 (nucleobase transporters), member 1; solute carrier family 23 (nucleobase transporters), member 2; solute carrier family 23 member 1; SVCT1; yolk sac permease-like molecule 3; YSPL3
Gene Symbols SLC23A1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC23A1. Recombinant protein control fragment (Product #RP-91177).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9963
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.