Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC23A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121468
This item is not returnable.
View return policy
Description
Antibody tested in Indirect ELISA.
The absorption of vitamin C into the body and its distribution to organs requires two sodium-dependent vitamin C transporters. This gene encodes one of the two required transporters and the encoded protein accounts for tissue-specific uptake of vitamin C. Previously, this gene had an official symbol of SLC23A1.The absorption of vitamin C into the body and its distribution to organs requires two sodium-dependent vitamin C transporters. This gene encodes one of the two required transporters and the encoded protein accounts for tissue-specific uptake of vitamin C. Previously, this gene had an official symbol of SLC23A1.
Specifications
| SLC23A2 | |
| Polyclonal | |
| Unconjugated | |
| SLC23A2 | |
| AI844736; hSVCT2; Kiaa0238; LOW QUALITY PROTEIN: solute carrier family 23 member 2; membrane transporter; mKIAA0238; mSVCT2; Na(+)/L-ascorbic acid transporter 2; NBTL1; Nucleobase transporter-like 1 protein; Slc23a1; SLC23A2; sodium-coupled ascorbic acid transporter 2; sodium-coupled ascorbic acid transporter SVCT2; sodium-dependent vitamin C transporter 2; sodium-dependent vitamin C transporter-2; solute carrier family 23 (ascorbic acid transporter), member 2; solute carrier family 23 (nucleobase transporters), member 1; solute carrier family 23 (nucleobase transporters), member 2; solute carrier family 23 member 2; SVCT2; SVCT-2; SVCT2-like protein; testicular secretory protein Li 48; yolk sac permease-like molecule 2; YSPL2; YSPL3 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 9962 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunocytochemistry | |
| 0.92 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9UGH3 | |
| SLC23A2 | |
| Recombinant Human Solute carrier family 23 member 2 protein (1-92aa). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction