Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC23A3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569285
Description
This target displays homology in the following species: Human: 100%.
SLC23A3 gene ontology annotations related to this gene include transmembrane transporter activity.
Specifications
SLC23A3 | |
Polyclonal | |
Unconjugated | |
SLC23A3 | |
E2-binding protein 3; E2BP3; HPC E2-binding protein 3; Na(+)/L-ascorbic acid transporter 3; SLC23A3; Sodium-dependent vitamin C transporter 3; solute carrier family 23 (nucleobase transporters), member 3; solute carrier family 23 member 3; solute carrier family 23, member 3; SVCT3; Yolk sac permease-like molecule 1; Yspl1; YSPL-1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
151295 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6PIS1 | |
SLC23A3 | |
synthetic peptide directed towards the N terminal region of human SLC23A3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction