Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC23A3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989922
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9922 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9922 Supplier Bioss Supplier No. BS20881R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC23A3 gene ontology annotations related to this gene include transmembrane transporter activity.

Specifications

Antigen SLC23A3
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH
Gene SLC23A3
Gene Accession No. Q6PIS1
Gene Alias E2-binding protein 3; E2BP3; HPC E2-binding protein 3; Na(+)/L-ascorbic acid transporter 3; SLC23A3; Sodium-dependent vitamin C transporter 3; solute carrier family 23 (nucleobase transporters), member 3; Solute carrier family 23 member 3; solute carrier family 23, member 3; SVCT3; Yspl1
Gene Symbols SLC23A3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from mouse SLC23A3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151295
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.