Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC24A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561020
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561020 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561020 Supplier Invitrogen™ Supplier No. PA561020
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CIHQCFERRT KGAGNMVNGL ANNAEIDDSS NCDATVVLLK KANFHRKASV IMVDELLSAY PHQLSF Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Plasma membrane sodium/calcium exchangers are an important component of intracellular calcium homeostasis and electrical conduction. Potassium-dependent sodium/calcium exchangers such as SLC24A3 are believed to transport 1 intracellular calcium and 1 potassium ion in exchange for 4 extracellular sodium ions (Kraev et al., 2001 [PubMed 11294880]).[supplied by OMIM, Mar 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC24A3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC24A3
Gene Accession No. Q9HC58
Gene Alias Na(+)/K(+)/Ca(2+)-exchange protein 3; NCKX3; Slc24a3; sodium calcium exchanger; sodium/potassium/calcium exchanger 3; solute carrier family 24 (sodium/potassium/calcium exchanger), member 3; solute carrier family 24 member 3
Gene Symbols SLC24A3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC24A3. Recombinant protein control fragment (Product #RP-99470).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57419
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.