Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC24A4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542518
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542518 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542518 Supplier Invitrogen™ Supplier No. PA542518
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PSLEKICERL HLSEDVAGAT FMAAGSSTPE LFASVIGVFI THGDVGVGTI Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

This gene encodes a member of the potassium-dependent sodium/calcium exchanger protein family. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC24A4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC24A4
Gene Accession No. Q8NFF2
Gene Alias A930002M03Rik; AI2A5; AW492432; K-dependent Na/Ca exchanger NCKX4; Na(+)/K(+)/Ca(2+)-exchange protein 4; NCKX4; SHEP6; SLC24A2; Slc24a4; Sodium/potassium/calcium exchanger 4; solute carrier family 24 (sodium/potassium/calcium exchanger), member 4; solute carrier family 24 member 4
Gene Symbols SLC24A4
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SLC24A4 (aa 124-173).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 123041
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.