Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC24A5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598527
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598527 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598527 Supplier Invitrogen™ Supplier No. PA598527
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC24A5 Polyclonal Antibody for Western Blot, ELISA

The SLC24A5 gene is a member of the potassium-dependent sodium/calcium exchanger family and encodes an intracellular membrane protein with 2 large hydrophilic loops and 2 sets of multiple transmembrane-spanning segments. Sequence variation in this gene has been associated with differences in skin pigmentation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC24A5
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SLC24A5
Gene Accession No. Q71RS6
Gene Alias F630045L20Rik; ion transporter JSX; JSX; K-independent Na+/Ca2+ exchanger JSX; Na(+)/K(+)/Ca(2+)-exchange protein 5; Na-Ca exchanger 5; NCKX5; NCX5; OCA6; oculocutaneous albinism 6 (autosomal recessive); SHEP4; Slc24a5; sodium/potassion/calcium exchanger 5; sodium/potassium/calcium exchanger 5; solute carrier family 24 (sodium/potassium/calcium exchanger), member 5; solute carrier family 24 member 5; solute carrier family 24, member 5
Gene Symbols SLC24A5
Host Species Rabbit
Immunogen Recombinant Human Sodium/potassium/calcium exchanger 5 protein (240-298AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283652
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.