Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC25A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA525403
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA525403 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA525403 Supplier Invitrogen™ Supplier No. PA525403
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC25A1 Polyclonal Antibody for Western Blot, IHC (P), Flow

The mitochondrial tricarboxylate transporter (also called citrate transport protein, or CTP) is responsible for the movement of citrate across the mitochondrial inner membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene SLC25A1
Gene Accession No. P53007
Gene Alias 1300019P08Rik; 2610100G11Rik; AI194714; Cic; Citrate transport protein; citrate transporter) member 1; clathrin heavy chain 2; clathrin heavy chain 2; uncharacterized protein LOC100053137; clathrin, heavy chain-like 1; CLTCL1; Ctp; D2L2AD; Dgsj; DiGeorge syndrome gene j; LOC100053137; mitochondrial tricarboxylate carrier; SEA; Slc20a3; SLC25A1; solute carrier family 20 (mitochondrial citrate transporter), member 3; solute carrier family 25 (mitochondrial carrier, citrate transporter), member 1; solute carrier family 25 (mitochondrial carrier; citrate transporter), member 1; solute carrier family 25 member 1; Tricarboxylate carrier protein; tricarboxylate transport protein, mitochondrial; txtp
Gene Symbols SLC25A1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 142-170 amino acids from the central region of human TXTP.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6576
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.