Learn More
Invitrogen™ SLC25A1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA585163
Description
Keep as concentrated solution. Predicted reactivity: Mouse (97%), Rat (96%), Xenopus laevis (88%), Pig (99%), Rhesus Monkey (100%), Bovine (97%). Positive Control: Huh-7, HepG2, mouse cerebellum, rat liver. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The mitochondrial tricarboxylate transporter (also called citrate transport protein, or CTP) is responsible for the movement of citrate across the mitochondrial inner membrane.
Specifications
SLC25A1 | |
Polyclonal | |
Unconjugated | |
SLC25A1 | |
1300019P08Rik; 2610100G11Rik; AI194714; Cic; Citrate transport protein; citrate transporter) member 1; clathrin heavy chain 2; clathrin heavy chain 2; uncharacterized protein LOC100053137; clathrin, heavy chain-like 1; CLTCL1; Ctp; D2L2AD; Dgsj; DiGeorge syndrome gene j; LOC100053137; mitochondrial tricarboxylate carrier; SEA; Slc20a3; SLC25A1; solute carrier family 20 (mitochondrial citrate transporter), member 3; solute carrier family 25 (mitochondrial carrier, citrate transporter), member 1; solute carrier family 25 (mitochondrial carrier; citrate transporter), member 1; solute carrier family 25 member 1; Tricarboxylate carrier protein; tricarboxylate transport protein, mitochondrial; txtp | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13358, 29743, 6576 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry, Western Blot | |
0.97 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P32089, P53007, Q8JZU2 | |
SLC25A1 | |
Recombinant protein encompassing a sequence within the center region of human SLC25A1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.