Learn More
SLC25A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 152351AP150UL

Description
This antibody specifically recognized the endogenous SLC25A1; no protein was detected in cells from subjects containing two alleles with truncating mutations with this antibody.
The mitochondrial tricarboxylate transporter (also called citrate transport protein, or CTP) is responsible for the movement of citrate across the mitochondrial inner membrane.Specifications
SLC25A1 | |
Polyclonal | |
Unconjugated | |
SLC25A1 | |
CIC, Citrate transport protein, CTP, SLC20A3, SLC25A1, Tricarboxylate carrier protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13358, 29743, 6576 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P32089, P53007, Q8JZU2 | |
SLC25A1 | |
SLC25A1 Fusion Protein Ag7352 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.