Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC25A11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596958
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596958 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596958 Supplier Invitrogen™ Supplier No. PA596958
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: MCF7, Raji, H460, Mouse brain, Rat brain; Cellular Location: Mitochondrion inner membrane, Multi-pass membrane protein Immunogen sequence: SAGAGGIDGK PRTSP.

The oxoglutarate/malate carrier transports 2-oxoglutarate across the inner membranes of mitochondria in an electroneutral exchange for malate or other dicarboxylic acids (summary by Iacobazzi et al., 1992 [PubMed 1457818]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A11
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.09 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SLC25A11
Gene Accession No. P97700, Q02978, Q9CR62
Gene Alias 2310022P18Rik; 2oxoc; 2-oxoglutarate carrier; 2-oxoglutarate/malate carrier; 2-oxoglutarate/malate carrier protein; fa07h09; Mitochondrial 2-oxoglutarate/malate carrier protein; OGC; OGCP; OMC; SLC20A4; SLC25A11; solute carrier family 20 (oxoglutarate carrier), member 4; solute carrier family 25 (mitochondrial carrier oxoglutarate carrier), member 11; solute carrier family 25 (mitochondrial carrier; oxoglutarate carrier), member 11; solute carrier family 25 member 11; wu:fa07h09; zgc:86898
Gene Symbols SLC25A11
Host Species Rabbit
Immunogen Synthetic Peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64201, 67863, 8402
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.