Learn More
SLC25A11 Polyclonal Antibody, Invitrogen™
Description
Recommended positive controls: Jurkat, Raji, rat kidney. Predicted reactivity: Mouse (96%), Rat (96%), Zebrafish (92%), Xenopus laevis (93%), Sheep (96%), Rhesus Monkey (100%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | SLC25A11 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | SLC25A11 |
| Gene Accession No. | P97700, Q02978, Q9CR62 |
| Gene Alias | 2310022P18Rik; 2oxoc; 2-oxoglutarate carrier; 2-oxoglutarate/malate carrier; 2-oxoglutarate/malate carrier protein; fa07h09; Mitochondrial 2-oxoglutarate/malate carrier protein; OGC; OGCP; OMC; SLC20A4; SLC25A11; solute carrier family 20 (oxoglutarate carrier), member 4; solute carrier family 25 (mitochondrial carrier oxoglutarate carrier), member 11; solute carrier family 25 (mitochondrial carrier; oxoglutarate carrier), member 11; solute carrier family 25 member 11; wu:fa07h09; zgc:86898 |
| Gene Symbols | SLC25A11 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.