Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC25A12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557234
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557234 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557234 Supplier Invitrogen™ Supplier No. PA557234
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAVKVQTTKR GDPHELRNIF LQYASTEVDG ERYMTPEDFV QRYLGLYNDP NSNPKIVQLL AGVADQTK Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 68%.

This gene encodes a calcium-binding mitochondrial carrier protein. The encoded protein localizes to the mitochondria and is involved in the exchange of aspartate for glutamate across the inner mitochondrial membrane. Polymorphisms in the gene encoding this protein may be associated with risk of autism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A12
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC25A12
Gene Accession No. O75746
Gene Alias 2610002D09Rik; AGC1; AI839531; araceli hiperlarga; ARALAR; Aralar1; B230107K20Rik; BB129864; calcium binding mitochondrial carrier superfamily member Aralar1; calcium-binding mitochondrial carrier protein Aralar1; Calcium-binding mitochondrial carrier protein Aralar1-like protein; mitochondrial aspartate glutamate carrier 1; RGD1561141; SLC25A12; solute carrier family 25 (aspartate/glutamate carrier), member 12; solute carrier family 25 (mitochondrial carrier, Aralar), member 12; solute carrier family 25 member 12; solute carrier family 25, member 12
Gene Symbols SLC25A12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC25A12. Recombinant protein control fragment (Product #RP-102663).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8604
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.