Learn More
Invitrogen™ SLC25A12 Recombinant Rabbit Monoclonal Antibody (10B12)

Description
SLC25A12 Recombinant Monoclonal Antibody for Western Blot, ELISA
Specifications
Specifications
| Antigen | SLC25A12 |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 10B12 |
| Concentration | 0.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | SLC25A12 |
| Gene Accession No. | O75746 |
| Gene Alias | 2610002D09Rik; AGC1; AI839531; araceli hiperlarga; ARALAR; Aralar1; B230107K20Rik; BB129864; calcium binding mitochondrial carrier superfamily member Aralar1; calcium-binding mitochondrial carrier protein Aralar1; Calcium-binding mitochondrial carrier protein Aralar1-like protein; mitochondrial aspartate glutamate carrier 1; RGD1561141; SLC25A12; solute carrier family 25 (aspartate/glutamate carrier), member 12; solute carrier family 25 (mitochondrial carrier, Aralar), member 12; solute carrier family 25 member 12; solute carrier family 25, member 12 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.