Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC25A13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553887
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553887 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553887 Supplier Invitrogen™ Supplier No. PA553887
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KVALTKRADP AELRTIFLKY ASIEKNGEFF MSPNDFVTRY LNIFGESQPN PKTVELLSGV VDQTKDGLIS Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

Citrin, also known as SLC25A13 (Solute carrier family 25 member 13), ARALAR2 or CTLN2, is a 675 amino acid multi-pass membrane protein that localizes to the inner membrane of the mitochondrion. Expressed in liver, pancreas, kidney, brain, heart and placenta, citrin functions as a calcium-dependent glutamate and aspartate carrier that is thought to play a role in the urea cycle. Citrin, a member of the mitochondrial carrier family, contains three Solcar repeats and four EF-hand domains through which it binds calcium. Defects in the gene encoding citrin are the cause of citrullinemia type 2 (CTLN2) and neonatal intrahepatic cholestasis due to citrin deficiency (NICCD). CTLN2 is an autosomal recessive disease that results from errors in the urea cycle and is characterized by neuropsychiatric symptoms such as loss of memory, seizures and coma. NICCD, a non-lethal disorder, occurs during infancy and is characterized by low birth weight, reduced bile flow, growth retardation and hepatic fibrosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A13
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Slc25a13
Gene Accession No. Q9UJS0
Gene Alias AI785475; ARALAR2; Calcium-binding mitochondrial carrier protein Aralar2; citrin; CTLN2; Ctrn; Mitochondrial aspartate glutamate carrier 2; RGD1565889; SLC25A13; solute carrier family 25 (aspartate/glutamate carrier), member 13; solute carrier family 25 (mitochondrial carrier, adenine nucleotide translocator), member 13; solute carrier family 25 member 13; solute carrier family 25, member 13; solute carrier family 25, member 13 (citrin)
Gene Symbols Slc25a13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC25A13. Recombinant protein control fragment (Product #RP-92588).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10165
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.