Learn More
Invitrogen™ SLC25A13 Polyclonal Antibody

Description
SLC25A13 Polyclonal Antibody for Western Blot, ELISA
Specifications
Specifications
| Antigen | SLC25A13 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.6 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Slc25a13 |
| Gene Accession No. | Q9UJS0 |
| Gene Alias | AI785475; ARALAR2; Calcium-binding mitochondrial carrier protein Aralar2; citrin; CTLN2; Ctrn; Mitochondrial aspartate glutamate carrier 2; RGD1565889; SLC25A13; solute carrier family 25 (aspartate/glutamate carrier), member 13; solute carrier family 25 (mitochondrial carrier, adenine nucleotide translocator), member 13; solute carrier family 25 member 13; solute carrier family 25, member 13; solute carrier family 25, member 13 (citrin) |
| Gene Symbols | Slc25a13 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.