Learn More
Invitrogen™ SLC25A13 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA588623
Description
Immunogen sequence: MAAAKVALTK RADPAELRTI FLKYASIEKN GEFFMSPNDF VTRYLNIFGE SQPNPKTVEL LSGVVDQTKD GLISFQEFVA FESVLCAPDA LFMVAFQLFD KAGKGEVTFE DVKQVFGQTT IHQHIPFNWD SEFVQLHFGK ERKRHLTYAE FTQFLLEIQL EHAKQAFVQR DNARTGRVTA IDFRDIMVTI RPHVLTPFVE ECLVAAAGGT TSHQVSFSYF NGFNSLLNNM ELIRKIYSTL AGTRKDVEVT KEEFVLAAQK FGQVTPMEVD ILFQLADLYE PRGRMTLADI ERIAPLEEGT; Positive Samples: MCF7, SW480, HepG2, HeLa ; Cellular Location: Mitochondrion inner membrane, Multi-pass membrane protein.
Citrin, also known as SLC25A13 (Solute carrier family 25 member 13), ARALAR2 or CTLN2, is a 675 amino acid multi-pass membrane protein that localizes to the inner membrane of the mitochondrion. Expressed in liver, pancreas, kidney, brain, heart and placenta, citrin functions as a calcium-dependent glutamate and aspartate carrier that is thought to play a role in the urea cycle. Citrin, a member of the mitochondrial carrier family, contains three Solcar repeats and four EF-hand domains through which it binds calcium. Defects in the gene encoding citrin are the cause of citrullinemia type 2 (CTLN2) and neonatal intrahepatic cholestasis due to citrin deficiency (NICCD). CTLN2 is an autosomal recessive disease that results from errors in the urea cycle and is characterized by neuropsychiatric symptoms such as loss of memory, seizures and coma. NICCD, a non-lethal disorder, occurs during infancy and is characterized by low birth weight, reduced bile flow, growth retardation and hepatic fibrosis.
Specifications
SLC25A13 | |
Polyclonal | |
Unconjugated | |
Slc25a13 | |
AI785475; ARALAR2; Calcium-binding mitochondrial carrier protein Aralar2; citrin; CTLN2; Ctrn; Mitochondrial aspartate glutamate carrier 2; RGD1565889; SLC25A13; solute carrier family 25 (aspartate/glutamate carrier), member 13; solute carrier family 25 (mitochondrial carrier, adenine nucleotide translocator), member 13; solute carrier family 25 member 13; solute carrier family 25, member 13; solute carrier family 25, member 13 (citrin) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10165, 362322, 50799 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Western Blot | |
0.82 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9QXX4, Q9UJS0 | |
Slc25a13 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human SLC25A13 (NP_0011536821). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.