Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC25A14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542431
Description
Peptide sequence: SIVAEFGTFP VDLTKTRLQV QGQSIDARFK EIKYRGMFHA LFRICKEEGV Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%; Yeast: 79%; Zebrafish: 100%.
Mitochondrial uncoupling proteins (UCP) are members of the larger family of mitochondrial anion carrier proteins (MACP). UCPs separate oxidative phosphorylation from ATP synthesis with energy dissipated as heat, also referred to as the mitochondrial proton leak. UCPs facilitate the transfer of anions from the inner to the outer mitochondrial membrane and the return transfer of protons from the outer to the inner mitochondrial membrane. They also reduce the mitochondrial membrane potential in mammalian cells. Tissue specificity occurs for the different UCPs and the exact methods of how UCPs transfer H+/OH- are not known. UCPs contain the three homologous protein domains of MACPs. This gene is widely expressed in many tissues with the greatest abundance in brain and testis. The gene product has an N-terminal hydrophobic domain that is not present in other UCPs. Two splice variants have been found for this gene.
Specifications
SLC25A14 | |
Polyclonal | |
Unconjugated | |
SLC25A14 | |
BMCP1; BMCP-1; brain mitochondrial carrier protein 1; MGC137654 protein; mitochondrial uncoupling protein 5; slc25a14; solute carrier family 25 (mitochondrial carrier, brain), member 14; solute carrier family 25 member 14; UCP 5; Ucp5; UCP5L; UCP5S; uncoupling protein 5; UNQ791/PRO1682; zgc:55596 | |
Rabbit | |
Protein A | |
RUO | |
9016 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O95258 | |
SLC25A14 | |
Synthetic peptide directed towards the N-terminal of human SLC25A14 (aa 51-100). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction