Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC25A15 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP220387
Description
SLC25A15 Polyclonal specifically detects SLC25A15 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SLC25A15 | |
Polyclonal | |
Unconjugated | |
0.1M Tris-Glycine (pH7), 20% Glycerol with 0.01% Thimerosal | |
D13S327, mitochondrial ornithine transporter 1, ORC1, ornithine transporter 1, ORNT1HHH, solute carrier family 25 (mitochondrial carrier; ornithine transporter) member15, Solute carrier family 25 member 15 | |
Rabbit | |
33 kDa | |
0.1 mL | |
Primary | |
Human. Species reactivity based on sequence identity: Mouse (95%), Chicken (81%), Bovine (95%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
0.71mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
Q9Y619 | |
SLC25A15 | |
Recombinant protein encompassing a sequence within the center region of human SLC25A15. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
10166 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction