Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC25A16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565169
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565169 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565169 Supplier Invitrogen™ Supplier No. PA565169
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RMQLGTVLPEF EKCLTMRDTM KYVYGHHGIR KGLY Highest antigen sequence identity to the following orthologs - mouse 89%, rat 91%.

SLC25A16 is a protein that contains three tandemly repeated mitochondrial carrier protein domains. The protein is localized in the inner membrane and facilitates the rapid transport and exchange of molecules between the cytosol and the mitochondrial matrix space. SLC25A16 gene has a possible role in Graves' disease.This gene encodes a protein that contains three tandemly repeated mitochondrial carrier protein domains. The encoded protein is localized in the inner membrane and facilitates the rapid transport and exchange of molecules between the cytosol and the mitochondrial matrix space. This gene has a possible role in Graves' disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A16
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC25A16
Gene Accession No. P16260
Gene Alias 3110021G18Rik; D10S105E; GDA; GDC; Graves disease autoantigen; Graves disease carrier protein; Graves disease carrier protein from bovine heart mitochondria; graves disease carrier protein homolog; HGT.1; hML7; MGC39851; mitochondrial carrier; human : Graves disease autoantigen; mitochondrial solute carrier protein homolog; ML7; SLC25A16; solute carrier family 25 (mitochondrial carrier), member 16; solute carrier family 25 (mitochondrial carrier, Graves disease autoantigen), member 16; solute carrier family 25 (mitochondrial carrier; Graves disease autoantigen), member 16; solute carrier family 25 member 16; solute carrier family 25, member 16; Unknown (protein for MGC:77742); wu:fc33c08; zgc:56187; zgc:77742
Gene Symbols SLC25A16
Host Species Rabbit
Immunogen Recombinant Human SLC25A16. Recombinant protein control fragment (Product #RP-97081).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8034
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.