Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC25A17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA526180
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA526180 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA526180 Supplier Invitrogen™ Supplier No. PA526180
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC25A17 Polyclonal Antibody for Western Blot, Flow

This protein encodes a selenoprotein, which contains a selenocysteine (Sec) residue at its active site. The selenocysteine is encoded by the UGA codon that normally signals translation termination. The 3' UTR of selenoprotein genes have a common stem-loop structure, the sec insertion sequence (SECIS), that is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. Studies suggest that this protein may regulate cytokine production, and thus play a key role in the control of the inflammatory response.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A17
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene SLC25A17
Gene Accession No. O43808, O70579
Gene Alias 34 kDa peroxisomal membrane protein; 34kDa; 47kDa; peroxisomal integral membrane protein 47; peroxisomal membrane protein PMP34; peroxisomal membrane protein, 34 kDa; Pmp34; Pmp35; Pmp47; SLC25A17; solute carrier family 25 (mitochondrial carrier, peroxisomal membrane protein), member 17; solute carrier family 25 (mitochondrial carrier; peroxisomal membrane protein, 34kDa), member 17; solute carrier family 25 member 17; solute carrier family 25, member 17
Gene Symbols SLC25A17
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 17-44 amino acids from the N-terminal region of human SLC25A17.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10478, 20524
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.