Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC25A18 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140123
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: NDFFRRLLME DGMQRNLKME M.
Involved in the transport of glutamate across the inner mitochondrial membrane. Glutamate is cotransported with H(+).
Specifications
SLC25A18 | |
Polyclonal | |
Unconjugated | |
SLC25A18 | |
1500015I14Rik; AW125787; Gc2; GC-2; glutamate carrier 2, mitochondrial; glutamate/H(+) symporter 2; Mitochondrial glutamate carrier 2; SLC25A18; solute carrier family 25 (glutamate carrier), member 18; solute carrier family 25 (mitochondrial carrier), member 18; solute carrier family 25 member 18 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83733 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H1K4 | |
SLC25A18 | |
Recombinant protein corresponding to Human SLC25A18. Recombinant protein control fragment (Product #RP-109149). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction