Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC25A19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542484
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542484 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542484 Supplier Invitrogen™ Supplier No. PA542484
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VHRGSVYDAR EFSVHFVCGG LAACMATLTV HPVDVLRTRF AAQGEPKVYN Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 93%; Yeast: 77%.

SLC25A19 encodes a mitochondrial protein that is a member of the solute carrier family. Although this protein was initially thought to be the mitochondrial deoxynucleotide carrier involved in the uptake of deoxynucleotides into the matrix of the mitochondria, further studies have demonstrated that this protein instead functions as the mitochondrial thiamine pyrophosphate carrier, which transports thiamine pyrophosphates into mitochondria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A19
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC25A19
Gene Accession No. Q9HC21
Gene Alias 2900089E13Rik; DNC; LOC504418 protein; MCPHA; mitochondrial deoxynucleotide carrier; mitochondrial thiamine pyrophosphate carrier; Mitochondrial uncoupling protein 1; MUP1; Slc25a19; solute carrier family 25 (mitochondrial deoxynucleotide carrier), member 19; solute carrier family 25 (mitochondrial thiamine pyrophosphate carrier), member 19; solute carrier family 25 member 19; solute carrier family 25, member 19; THMD3; THMD4; TPC
Gene Symbols SLC25A19
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human SLC25A19.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60386
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.