Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC25A2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570702
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%.
Located between the protocadherin beta and gamma gene clusters on chromosome 5, this intronless gene encodes a protein that is highly similar to an ornithine transporter localized in the mitochondrial inner membrane. The encoded protein most likely plays a role in metabolism as a mitochondrial transport protein.
Specifications
SLC25A2 | |
Polyclonal | |
Unconjugated | |
SLC25A2 | |
mitochondrial ornithine transporter; mitochondrial ornithine transporter 2; ORC2; ornithine transporter 2; Ornt2; SLC25A2; solute carrier family 25 (mitochondrial carrier, ornithine transporter) member 2; solute carrier family 25 (mitochondrial carrier; ornithine transporter) member 2; solute carrier family 25 member 2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
83884 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BXI2 | |
SLC25A2 | |
synthetic peptide directed towards the C-terminal region of human SLC25A2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction