Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC25A21 Monoclonal Antibody (2C9)
GREENER_CHOICE

Catalog No. PIMA522830
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522830 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522830 Supplier Invitrogen™ Supplier No. MA522830
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: VVKTRFQIQR CATDPNSYKS LVDSFRMIFQ MEGLFGFYKG ILPPILAETP KRAVKFFTFE QYKK.

SLC25A21 is a homolog of the S. cerevisiae ODC proteins, mitochondrial carriers that transport C5-C7 oxodicarboxylates across inner mitochondrial membranes. One of the species transported by ODC is 2-oxoadipate, a common intermediate in the catabolism of lysine, tryptophan, and hydroxylysine in mammals. Within mitochondria, 2-oxoadipate is converted into acetyl-CoA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A21
Applications ELISA
Classification Monoclonal
Clone 2C9
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene SLC25A21
Gene Accession No. Q9BQT8
Gene Alias 9930033G19Rik; A630030I10; BB158148; MGC126570; mitochondrial 2-oxodicarboxylate carrier; mitochondrial solute carrier family 25 member 21; Odc; Odc1; oxodicarboxylate carrier; SLC25A21; solute carrier family 25 (mitochondrial oxoadipate carrier), member 21; solute carrier family 25 (mitochondrial oxodicarboxylate carrier), member 21; solute carrier family 25 member 21; zgc:153387
Gene Symbols SLC25A21
Host Species Mouse
Immunogen SLC25A21 (NP_085134.1, 36 a.a. ~ 99 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89874
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.