Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC25A22 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530822
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530822 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530822 Supplier Invitrogen™ Supplier No. PA530822
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A431, mouse brain. Predicted reactivity: Mouse (94%), Chicken (81%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The SLC25 gene family encodes mitochondrial carriers that transport a variety of metabolites across the inner mitochondrial membrane (Palmieri, 2004 ). SLC25A22, also known as GC1, is 1 of the 2 mitochondrial glutamate/H+ symporters, the other being SLC25A18.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A22
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Slc25a22
Gene Accession No. Q9D6M3, Q9H936
Gene Alias 1300006L01Rik; AI060884; EIEE3; FLJ13044; GC1; GC-1; Glutamate/H(+) symporter 1; mitochondrial glutamate carrier 1; NET44; RGD1307826; Slc25a22; solute carrier family 25 (mitochondrial carrier, glutamate), member 22; solute carrier family 25 (mitochondrial carrier: glutamate), member 22; solute carrier family 25 member 22
Gene Symbols Slc25a22
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SLC25A22. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 68267, 79751
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.