Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC25A22 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542492
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542492 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542492 Supplier Invitrogen™ Supplier No. PA542492
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LQDAGRIAAQ RKILAAQGQL SAQGGAQPSV EAPAAPRPTA TQLTRDLLRS Sequence homology: Cow: 86%; Guinea Pig: 92%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 93%; Rat: 86%; Zebrafish: 100%.

The SLC25 gene family encodes mitochondrial carriers that transport a variety of metabolites across the inner mitochondrial membrane (Palmieri, 2004 ). SLC25A22, also known as GC1, is 1 of the 2 mitochondrial glutamate/H+ symporters, the other being SLC25A18.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A22
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Slc25a22
Gene Accession No. Q9H936
Gene Alias 1300006L01Rik; AI060884; EIEE3; FLJ13044; GC1; GC-1; Glutamate/H(+) symporter 1; mitochondrial glutamate carrier 1; NET44; RGD1307826; Slc25a22; solute carrier family 25 (mitochondrial carrier, glutamate), member 22; solute carrier family 25 (mitochondrial carrier: glutamate), member 22; solute carrier family 25 member 22
Gene Symbols Slc25a22
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SLC25A22 (aa 131-180).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79751
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.