Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC25A23 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584033
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584033 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584033 Supplier Invitrogen™ Supplier No. PA584033
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GRLFEELDSN KDGRVDVHEL RQGLARLGGG NPDPGAQQGI SSEGDADPDG GLD Highest antigen sequence indentity to the following orthologs: Mouse - 85%, Rat - 83%.

Calcium-dependent mitochondrial solute carrier. Mitochondrial solute carriers shuttle metabolites, nucleotides, and cofactors through the mitochondrial inner membrane (PubMed:15123600). May act as a ATP-Mg/Pi exchanger that mediates the transport of Mg-ATP in exchange for phosphate, catalyzing the net uptake or efflux of adenine nucleotides into or from the mitochondria (PubMed:15123600). Acts as a regulator of mitochondrial calcium uptake via interaction with MCU and MICU1 (PubMed:24430870). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A23
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC25A23
Gene Accession No. Q9BV35
Gene Alias 2310067G05Rik; APC2; calcium-binding mitochondrial carrier protein SCaMC-3; MCSC2; mitochondrial ATP-Mg/Pi carrier protein 2; Mitochondrial Ca(2+)-dependent solute carrier protein 2; mitochondrial Ca2+-dependent solute carrier protein 2; phosphate carrier), member 23; SCAMC3; SCaMC-3; short calcium-binding mitochondrial carrier 3; Slc25a23; sma; Small calcium-binding mitochondrial carrier protein 3; solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 23; solute carrier family 25 member 23
Gene Symbols SLC25A23
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC25A23. Recombinant protein control fragment (Product #RP-101261).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79085
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.