Learn More
Invitrogen™ SLC25A24 Polyclonal Antibody

Description
Immunogen sequence: MLRWLRDFVL PTAACQDAEQ PTRYETLFQA LDRNGDGVVD IGELQEGLRN LGIPLGQDAE EKIFTTGDVN KDGKLDFEEF MKYLKDHEKK MKLAFKSLDK NNDGKIEASE IVQSLQTLGL TISEQQAELI LQSIDVDGTM TVDWNEWRDY FLFNPVTDIE EIIRFWKHST GIDIGDSLTI PDEFTEDEKK SGQWWRQLLA; Positive Samples: BxPC-3, U-87MG, HT-29, 293T, NIH/3T3, Mouse spleen, Rat spleen; Cellular Location: Mitochondrion inner membrane, Multi-pass membrane protein.
Specifications
Specifications
| Antigen | SLC25A24 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.35 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | SLC25A24 |
| Gene Accession No. | Q6NUK1, Q8BMD8 |
| Gene Alias | 2610016M12Rik; APC1; Calcium-binding mitochondrial carrier protein SCaMC-1; calcium-binding mitochondrial carrier protein SCaMC-1-like; Calcium-binding mitochondrial carrier protein SCaMC-1-like protein; calcium-binding transporter; DKFZp586G0123; EFINAL; MCSC1; Mitochondrial ATP-Mg/Pi carrier protein 1; mitochondrial Ca(2+)-dependent solute carrier protein 1; Peroxisomal Ca(2+)-dependent solute carrier; peroxisomal Ca-dependent solute carrier; RP11-356N1.3; scamc1; SCAMC-1; short calcium-binding mitochondrial carrier 1; si:ch211-63o20.9; similar to mitochondrial solute carrier; Slc25a24; Small calcium-binding mitochondrial carrier protein 1; solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 24; solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 24; solute carrier family 25 member 24; zgc:92470 |
| Gene Symbols | SLC25A24 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.