Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC25A24 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598508
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598508 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598508 Supplier Invitrogen™ Supplier No. PA598508
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC25A24 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

SLC25A24 is a calcium-dependent mitochondrial solute carrier. Mitochondrial solute carriers shuttle metabolites, nucleotides, and cofactors through the mitochondrial inner membrane. It may act as a ATP-Mg/Pi exchanger that mediates the transport of Mg-ATP in exchange for phosphate, catalyzing the net uptake or efflux of adenine nucleotides into or from the mitochondria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A24
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SLC25A24
Gene Accession No. Q6NUK1, Q8BMD8
Gene Alias 2610016M12Rik; APC1; Calcium-binding mitochondrial carrier protein SCaMC-1; calcium-binding mitochondrial carrier protein SCaMC-1-like; Calcium-binding mitochondrial carrier protein SCaMC-1-like protein; calcium-binding transporter; DKFZp586G0123; EFINAL; MCSC1; Mitochondrial ATP-Mg/Pi carrier protein 1; mitochondrial Ca(2+)-dependent solute carrier protein 1; Peroxisomal Ca(2+)-dependent solute carrier; peroxisomal Ca-dependent solute carrier; RP11-356N1.3; scamc1; SCAMC-1; short calcium-binding mitochondrial carrier 1; si:ch211-63o20.9; similar to mitochondrial solute carrier; Slc25a24; Small calcium-binding mitochondrial carrier protein 1; solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 24; solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 24; solute carrier family 25 member 24; zgc:92470
Gene Symbols SLC25A24
Host Species Rabbit
Immunogen Recombinant Human Calcium-binding mitochondrial carrier protein SCaMC-1 protein (1-197AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 229731, 29957, 310791
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.