Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC25A25 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA582724
Description
Immunogen sequence: LRLVFKSLDK KNDGRIDAQE IMQSLRDLGV KISEQQAEKI LKSMDKNGTM TIDWNEWRDY HLLHPVENIP EIILYWKHST IFDVGENLT.
Calcium-dependent mitochondrial solute carrier. Mitochondrial solute carriers shuttle metabolites, nucleotides, and cofactors through the mitochondrial inner membrane. May act as a ATP-Mg/Pi exchanger that mediates the transport of Mg-ATP in exchange for phosphate, catalyzing the net uptake or efflux of adenine nucleotides into or from the mitochondria.
Specifications
SLC25A25 | |
Polyclonal | |
Unconjugated | |
SLC25A25 | |
1110030N17Rik; APC3; ATP-Mg/Pi carrier; calcium-binding mitochondrial carrier protein SCaMC-2; Kiaa1896; LOW QUALITY PROTEIN: calcium-binding mitochondrial carrier protein SCaMC-2; MCSC; MCSC3; Mitochondrial ATP-Mg/Pi carrier protein; mitochondrial ATP-Mg/Pi carrier protein 3; Mitochondrial Ca(2+)-dependent solute carrier protein 3; mitochondrial Ca2+-dependent solute carrier; mKIAA1896; Pcscl; peroxisomal Ca(2+)-dependent solute carrier-like protein; peroxisomal Ca-dependent solute carrier-like protein; Scamc2; SCAMC-2; short calcium-binding mitochondrial carrier 2; SLC25A25; small calcium-binding mitochondrial carrier 2; small calcium-binding mitochondrial carrier protein 2; solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 25; solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 25; solute carrier family 25 member 25; Unknown (protein for MGC:127973); UNQ549/PRO1106 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114789, 227731, 246771 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A2ASZ8, Q6KCM7, Q8K3P6 | |
SLC25A25 | |
Recombinant protein corresponding to Human SLC25A25. Recombinant protein control fragment (Product #RP-93011). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction