Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC25A25 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113047
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
PIPA5113047 100 μg
1 options

Catalog No. PIPA5113047

Supplier: Invitrogen™ PA5113047

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC25A25 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Calcium-dependent mitochondrial solute carrier. Mitochondrial solute carriers shuttle metabolites, nucleotides, and cofactors through the mitochondrial inner membrane. May act as a ATP-Mg/Pi exchanger that mediates the transport of Mg-ATP in exchange for phosphate, catalyzing the net uptake or efflux of adenine nucleotides into or from the mitochondria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC25A25
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SLC25A25
Gene Accession No. A2ASZ8, Q6KCM7
Gene Alias 1110030N17Rik; APC3; ATP-Mg/Pi carrier; calcium-binding mitochondrial carrier protein SCaMC-2; Kiaa1896; LOW QUALITY PROTEIN: calcium-binding mitochondrial carrier protein SCaMC-2; MCSC; MCSC3; Mitochondrial ATP-Mg/Pi carrier protein; mitochondrial ATP-Mg/Pi carrier protein 3; Mitochondrial Ca(2+)-dependent solute carrier protein 3; mitochondrial Ca2+-dependent solute carrier; mKIAA1896; Pcscl; peroxisomal Ca(2+)-dependent solute carrier-like protein; peroxisomal Ca-dependent solute carrier-like protein; Scamc2; SCAMC-2; short calcium-binding mitochondrial carrier 2; SLC25A25; small calcium-binding mitochondrial carrier 2; small calcium-binding mitochondrial carrier protein 2; solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 25; solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 25; solute carrier family 25 member 25; Unknown (protein for MGC:127973); UNQ549/PRO1106
Gene Symbols SLC25A25
Host Species Rabbit
Immunogen Recombinant Human Calcium-binding mitochondrial carrier protein SCaMC-2 protein (1-189AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114789, 227731
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.